
Sodium Cyclamate

Synonym(s):
Sodium N-cyclohexylsulfamate, N-Cyclohexylsulfamic acid sodium salt, Cyclamic acid sodium salt, Cyclohexanesulfamic acid sodium salt, Sodium cyclamate, Sodium cyclohexanesulfamate
Linear Formula:
C6H11NHSO3Na
CAS Number:
Molecular Weight:
201.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing