Silver nitrate solution
Synonym(s):
Silver nitrate solution
Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Package Size
Reaction Type
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing