
Rivastigmine Related Compound F

Synonym(s):
3-Vinylphenyl ethyl(methyl)carbamate
Empirical Formula (Hill Notation):
C12H15NO2
CAS Number:
Molecular Weight:
205.25
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2