Skip to Content
Merck
(R)-(−)-2-Chloro-1-phenylethanol 97%

(R)-(−)-2-Chloro-1-phenylethanol

(R)-(−)-2-Chloro-1-phenylethanol 97%
Synonym(s):
(R)-(−)-α-(Chloromethyl)benzenemethanol
Linear Formula:
C6H5CH(CH2Cl)OH
CAS Number:
Molecular Weight:
156.61

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing