![Povidone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/142/228/0e1e7bd7-9aa1-4b0c-b64f-8d1fed4f06cc/640/0e1e7bd7-9aa1-4b0c-b64f-8d1fed4f06cc.png)
Povidone
![Povidone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/142/228/0e1e7bd7-9aa1-4b0c-b64f-8d1fed4f06cc/640/0e1e7bd7-9aa1-4b0c-b64f-8d1fed4f06cc.png)
Synonym(s):
Polyvinylpyrrolidone, PVP, Polyvidone, Povidone
Linear Formula:
(C6H9NO)n
CAS Number:
Products
Product Category
Brand
Feature
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4