Skip to Content
CO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phenylglyoxylic acid
Synonym(s):
α-Oxophenylacetic acid, Benzoylformic acid
Linear Formula:
C
6
H
5
COCOOH
CAS Number:
611-73-4
Molecular Weight:
150.13
EC Number:
210-278-7
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Purum (1)
certified reference material (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
B13055
Phenylglyoxylic acid,
97%
Expand
View Pricing
PHR9245
Phenylglyoxylic acid,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
78610
Phenylglyoxylic acid,
purum, ≥98.0% (T)
Expand
View Pricing