
Phellopterin

Empirical Formula (Hill Notation):
C17H16O5
CAS Number:
Molecular Weight:
300.31
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing