
Paroxetine Related Compound F

Synonym(s):
N-Methylparoxetine, (3S-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-1-methylpiperidine
Empirical Formula (Hill Notation):
C20H22FNO3
CAS Number:
Molecular Weight:
343.39
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2