
Pantoprazole sodium

Synonym(s):
Pantoprazole sodium sesquihydrate
Empirical Formula (Hill Notation):
C16H14F2N3NaO4S · 1.5H2O
CAS Number:
Molecular Weight:
432.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2