Skip to Content
Merck
ortho-Phosphoric acid 88% for analysis EMPARTA® ACS

ortho-Phosphoric acid 88%

ortho-Phosphoric acid 88% for analysis EMPARTA® ACS
Synonym(s):
Phosphoric acid, Orthophosphoric acid
Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing