
Orciprenaline for system suitability

Synonym(s):
Metaproterenol hemisulfate salt
Linear Formula:
C11H17NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
260.30
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing