Skip to Content
Merck
n-Undecane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

n-Undecane

n-Undecane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
n-Undecane, Undecane, n-Undecane, Hendecane
Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique