![Nuclease micrococcal from Staphylococcus aureus With reaction buffer and BSA solution](/deepweb/assets/sigmaaldrich/product/images/225/425/42afc676-19ad-42f2-a94e-89f9eef42c29/640/42afc676-19ad-42f2-a94e-89f9eef42c29.jpg)
Nuclease micrococcal from Staphylococcus aureus
![Nuclease micrococcal from Staphylococcus aureus With reaction buffer and BSA solution](/deepweb/assets/sigmaaldrich/product/images/225/425/42afc676-19ad-42f2-a94e-89f9eef42c29/640/42afc676-19ad-42f2-a94e-89f9eef42c29.jpg)
Synonym(s):
Endonuclease micrococcal, MNase, Micrococcal endonuclease, Nuclease micrococcal from Staphylococcus aureus
CAS Number:
Products
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-3 of 3