
Norgestimate

Synonym(s):
(17α)-17-(Acetyloxy)-13-ethyl-18,19-dinorpregn-4-en-20-yn-3-one 3-oxime, Dexnorgestrel acetime, Norgestimate
Empirical Formula (Hill Notation):
C23H31NO3
CAS Number:
Molecular Weight:
369.50
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing