
Nicotine Related Compound G

Synonym(s):
Anabasine, (S)-3-(Piperidin-2-yl)pyridine
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2