![Naphthol AS-D Chloroacetate](/deepweb/assets/sigmaaldrich/product/structures/156/930/28763aec-08ae-46e4-a4b4-b255067fb5bd/640/28763aec-08ae-46e4-a4b4-b255067fb5bd.png)
Naphthol AS-D chloroacetate
![Naphthol AS-D Chloroacetate](/deepweb/assets/sigmaaldrich/product/structures/156/930/28763aec-08ae-46e4-a4b4-b255067fb5bd/640/28763aec-08ae-46e4-a4b4-b255067fb5bd.png)
Synonym(s):
3-Hydroxy-2-naphthoic-o-toluidide chloroacetate, Naphthol AS-D chloroacetate
Empirical Formula (Hill Notation):
C20H16ClNO3
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing