Skip to Content
Merck
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly

N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly

N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly
Synonym(s):
FAPGG, N-[3-(2-Furyl)acryloyl]-L-phenylalanyl-glycyl-glycine
Empirical Formula (Hill Notation):
C20H21N3O6
CAS Number:
Molecular Weight:
399.40

Products

Product Category
Brand
Formula Weight
Gene Alias
Showing 1-1 of 1
Compare
Product Number
Description
Pricing