![Mycophenolate Sodium pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/282/255/d272feb5-5d59-49a2-86c1-f72729887e72/640/d272feb5-5d59-49a2-86c1-f72729887e72.jpg)
Mycophenolate sodium
![Mycophenolate Sodium pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/282/255/d272feb5-5d59-49a2-86c1-f72729887e72/640/d272feb5-5d59-49a2-86c1-f72729887e72.jpg)
Synonym(s):
Mycophenolic acid monosodium salt, Sodium mycophenolate
Empirical Formula (Hill Notation):
C17H19NaO6
CAS Number:
Molecular Weight:
342.32
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2