![Morphine-6-β-D-glucuronide solution 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/480/481/9f61066c-7082-452c-91ce-5f6abd7f139e/640/9f61066c-7082-452c-91ce-5f6abd7f139e.png)
Morphine-6-β-D-glucuronide solution
![Morphine-6-β-D-glucuronide solution 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/480/481/9f61066c-7082-452c-91ce-5f6abd7f139e/640/9f61066c-7082-452c-91ce-5f6abd7f139e.png)
Empirical Formula (Hill Notation):
C23H27NO9
CAS Number:
Molecular Weight:
461.46
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing