
Molindone hydrochloride

Synonym(s):
3-Ethyl-1,5,6,7-tetrahydro-2-methyl-5-(4-morpholinylmethyl)-4H-indol-4-one hydrochloride, Molindone hydrochloride
Empirical Formula (Hill Notation):
C16H24N2O2 · HCl
CAS Number:
Molecular Weight:
312.83
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2