
Micro particles based on melamine resin, carboxylate-modified, FITC-marked

Synonym(s):
Melamine-formaldehyde resin
Products
Product Category
Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing