Skip to Content
Merck
Mibolerone ≥98% (HPLC)

Mibolerone

Mibolerone ≥98% (HPLC)
Synonym(s):
(7α,17β)-17-Hydroxy-7,17-dimethylestr-4-en-3-one, 7α,17α-Dimethyl-19-nortestosterone
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing