Skip to Content
Merck
Metronidazole certified reference material, TraceCERT®

Metronidazole

Metronidazole certified reference material, TraceCERT®
Synonym(s):
2-Methyl-5-nitroimidazole-1-ethanol
Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique