
Metoclopramide impurity E

Synonym(s):
N,N-Diethylethylenediamine, 2-Diethylaminoethylamine, N,N-Diethylethylenediamine
Linear Formula:
(C2H5)2NCH2CH2NH2
CAS Number:
Molecular Weight:
116.20
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2