Skip to Content
Merck
Methyl methacrylate SAJ first grade, ≥99.0%

Methyl methacrylate

Methyl methacrylate SAJ first grade, ≥99.0%
Synonym(s):
Methyl methacrylate
Linear Formula:
CH2=C(CH3)COOCH3
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Monomer Type
Reference Material Type
Special Grade