
MES-d13

Synonym(s):
4-Morpholineethanesulfonic acid-d13 monohydrate
Empirical Formula (Hill Notation):
C6D13NO4S
Molecular Weight:
208.32
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing