
Macelignan

Synonym(s):
(+)-Anwulignan, 4-[(2S,3R)-4-(1,3-Benzodioxol-5-yl)-2,3-dimethylbutyl]-2-methoxy-phenol, Anwulignan
Empirical Formula (Hill Notation):
C20H24O4
CAS Number:
Molecular Weight:
328.40
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing