
LuminoCt® SYBR® Green qPCR ReadyMix™

Synonym(s):
LuminoCt®
Products
Product Category
Brand
Color
Physical Form
Number of Reactions
Compatibility
Quality Segment
dNTPs
Hot Start
Application
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing