Lipopolysaccharides from Salmonella enterica serotype typhimurium
Synonym(s):
LPS
Products
Product Category
Brand
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing