
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

Synonym(s):
Bandeirea simplicifolia agglutinin, BS-I
Products
Brand
Biological Source
Physical Form
Chemical Composition
Conjugate
Quality Segment
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing