
L-Alanine tert-butyl ester hydrochloride

Linear Formula:
CH3CH(NH2)COOC(CH3)3 · HCl
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing