
Hydroxybufuralol maleate salt

Empirical Formula (Hill Notation):
C16H23NO3 · C4H4O4
Molecular Weight:
393.43
Products
Brand
Color
Formula Weight
Physical Form
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing