
Hydrochloric acid solution

Synonym(s):
Hydrochloric acid solution, Hydrogen chloride solution
Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Agency
Volume (ml)
Quality Segment
Package Size
Reaction Type
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing