![Haloxyfop certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/187/735/1508155f-c17f-426b-be3d-89419fece4cb/640/1508155f-c17f-426b-be3d-89419fece4cb.png)
Haloxyfop
![Haloxyfop certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/187/735/1508155f-c17f-426b-be3d-89419fece4cb/640/1508155f-c17f-426b-be3d-89419fece4cb.png)
Synonym(s):
2-[4-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]propanoic acid, Haloxyfop acid
Empirical Formula (Hill Notation):
C15H11ClF3NO4
CAS Number:
Molecular Weight:
361.70
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2