
Gabapentin Related Compound B

Synonym(s):
(1-Cyanocyclohexyl)acetic acid, 1-Cyanocyclohexaneacetic acid
Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2