
Fluazifop

Synonym(s):
2-[4-(5-Trifluoromethyl-2-pyridyloxy)phenoxy]propionic acid
Empirical Formula (Hill Notation):
C15H12F3NO4
CAS Number:
Molecular Weight:
327.26
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing