Skip to Content
Merck
Farnesal, mixture of isomers ≥85%

Farnesal, mixture of isomers

Farnesal, mixture of isomers ≥85%
Synonym(s):
3,7,11-Trimethyl-2,6,10-dodecatrienal
Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing