
Famotidine for system suitability

Synonym(s):
Famotidine, N′-(Aminosulfonyl)-3-([2-(diaminomethyleneamino)-4-thiazolyl]methylthio)propanamidine
Empirical Formula (Hill Notation):
C8H15N7O2S3
CAS Number:
Molecular Weight:
337.45
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing