
Exemestane for peak identification

Synonym(s):
Exemestane, 6-Methyleneandrosta-1,4-diene-3,17-dione
Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Products
Formula Weight
Gene Alias
Manufacturer
Quality Segment
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing