![Eugenol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/199/694/3186859d-b25f-4a24-bce1-9969e4323749/640/3186859d-b25f-4a24-bce1-9969e4323749.png)
Eugenol
![Eugenol European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/199/694/3186859d-b25f-4a24-bce1-9969e4323749/640/3186859d-b25f-4a24-bce1-9969e4323749.png)
Synonym(s):
2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol, Eugenol
Linear Formula:
4-(H2C=CHCH2)C6H3-2-(OCH3)OH
CAS Number:
Molecular Weight:
164.20
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-10 of 10
Compare
Product Number
Description
Pricing