
Ethyl 4-amino-1-piperidinecarboxylate

Synonym(s):
1-Carbethoxy-4-aminopiperidine
Empirical Formula (Hill Notation):
C8H16N2O2
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing