
Ethiprole

Synonym(s):
5-Amino-1-(2,6-dichloro-α,α,α-trifluoro-p-tolyl)-4-(ethylsulfinyl)pyrazole-3-carbonitrile
Empirical Formula (Hill Notation):
C13H9Cl2F3N4OS
CAS Number:
Molecular Weight:
397.20
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2