
Ethanol-d6

Synonym(s):
Ethyl alcohol-d6, Ethyl-d5 alcohol-d, Hexadeuteroethanol
Linear Formula:
CD3CD2OD
CAS Number:
Molecular Weight:
52.11
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing