
Equilin

Synonym(s):
1,3,5(10),7-Estratetraen-3-ol-17-one, 3-Hydroxy-1,3,5(10),7-estratetraen-17-one, 7-Dehydroestrone
Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing