Skip to Content
Merck
ECO BRIJ® O20 average Mn ~1,150

ECO BRIJ® O20

ECO BRIJ® O20 average Mn ~1,150
Synonym(s):
BRIJ® O20, Polyoxyethylene (20) oleyl ether
Linear Formula:
C18H35(OCH2CH2)nOH, n~20
CAS Number:
Molecular Weight:
1149.53

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing