Skip to Content
Merck
Diethylene glycol SAJ first grade, ≥98.0%

Diethylene glycol

Diethylene glycol SAJ first grade, ≥98.0%
Synonym(s):
2,2′-Oxydiethanol, 2-Hydroxyethyl ether, Bis(2-hydroxyethyl) ether, Diglycol
Linear Formula:
(HOCH2CH2)2O
CAS Number:
Molecular Weight:
106.12
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Manufacturer
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique