
Desbenzyl donepezil hydrochloride

Synonym(s):
5,6-Dimethoxy-2-(piperidin-4-ylmethyl)indan-1-one hydrochloride
Empirical Formula (Hill Notation):
C17H23NO3 · HCl
CAS Number:
Molecular Weight:
325.83
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2