
Dabco® 33-LV

Synonym(s):
1,4-Diazabicyclo[2.2.2]octane solution
Empirical Formula (Hill Notation):
C6H12N2
CAS Number:
Molecular Weight:
112.17
Products
Product Category
Brand
Formula Weight
Quality Segment
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing