
(+)-3-(Trifluoroacetyl)camphor

Synonym(s):
3-(Trifluoroacetyl)-D-camphor
Empirical Formula (Hill Notation):
C12H15F3O2
Products
Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing