Skip to Content
Merck
Crotamiton British Pharmacopoeia (BP) Reference Standard

Crotamiton

Crotamiton British Pharmacopoeia (BP) Reference Standard
Synonym(s):
N-Ethyl-o-crotonotoluidide, N-Ethyl-N-(2-methylphenyl)-2-butenamide, Crotamiton
Linear Formula:
CH3CH=CHCON(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
203.28
EC Number:

Products

Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade